Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, Purified (Diammonium Hydrogen Phosphate), Reagents

Catalog No. p-200021012
Click to view available options
Quantity:
100 g
500 g

Ammonium Phosphate Dibasic, Purified (Diammonium Hydrogen Phosphate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym Diammonium Hydrogen Phosphate
IUPAC Name phosphoric acid diamine
SMILES N.N.OP(O)(O)=O

Specifications

Melting Point 155°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.619
Chemical Name or Material Ammonium Phosphate Dibasic, Purified (Diammonium Hydrogen Phosphate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.