Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Dibasic, puriss. p.a., Honeywell Fluka™

Click to view available options
Quantity:
50 g
250 g
1 kg
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
N.N.OP(O)(O)=O
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
N.N.OP(O)(O)=O

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Quantity 50 g
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
Formula Weight 132.06g/mol
Percent Purity ≥99% (T)
Grade puriss. p.a.
Chemical Name or Material Ammonium Phosphate Dibasic

EINECSNumber : 231-987-8