Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate, Monobasic, Crystal, ACS, 98%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7722-76-1
7722-76-1
Grade ACS
CAS Max % 1
Percent Purity 0.98
Calcium (Ca) 1e-05
Chloride 5 ppm
Heavy Metals 5 ppm
Insoluble Matter 5e-05
Iron (Fe) 1e-05
Magnesium (Mg) 5e-06
Nitrate 1e-05
Packaging Poly Bottle
Potassium (K) 5e-05
Sodium (Na) 5e-05
Sulfate 0.0001
pH 3.8 to 4.4
Quantity 2.5 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.