Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate, Monobasic, FCC, 96-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7722-76-1
H6NO4P
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate
[NH4+].OP(O)([O-])=O
7722-76-1
H6NO4P
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate
[NH4+].OP(O)([O-])=O

AM183, 7722-76-1, NH4H2PO4

Spectrum™ Ammonium Phosphate, Monobasic, FCC is approved for usage in food and beverage applications as an acidity regulator and firming agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 96 to 102%
Fluoride 10 mg/kg
Lead (Pb) 4 mg/kg
Packaging Poly Bottle
Quantity 500 g