Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Monobasic, Purified (Ammonium Dihydrogen Phosphate), Reagents

Catalog No. 01337486
Click to view available options
Quantity:
100 g
500 g

Ammonium Phosphate Monobasic, Purified (Ammonium Dihydrogen Phosphate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym Ammonium dihydrogen phosphate, Monoammonium phosphate
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Boiling Point 197°C
Melting Point 190°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.803
Chemical Name or Material Ammonium Phosphate Monobasic, Purified (Ammonium Dihydrogen Phosphate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.