Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Sodium Phosphate Dibasic Tetrahydrate, Honeywell Fluka™
Click to view available options
Quantity:
500g
1kg
6 x 1kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | NaNH4HPO4 · 4H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 209.07 |
Percent Purity | ≥99% |
Grade | puriss. |
Quantity | 6 x 1kg |
Chemical Name or Material | Ammonium Sodium Phosphate Dibasic Tetrahydrate |