Learn More
Ammonium sulfate, 98+%, Thermo Scientific Chemicals
Description
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.
Solubility
Soluble in water. Insoluble in alcohol and acetone.
Notes
Incompatible with various substances like highly reactive oxidizing agents and alkalis.
Specifications
Melting Point | ∼280°C (decomposition) |
Quantity | 5000 g |
Linear Formula | (NH4)2SO4 |
Merck Index | 14,555 |
Solubility Information | Soluble in water. Insoluble in alcohol and acetone. |
Formula Weight | 132.14 |
Percent Purity | ≥98% |
Density | 1.769 |
Chemical Name or Material | Ammonium sulfate |
Safety and Handling
EINECSNumber | 231-984-1 |
RTECSNumber | BS4500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.