Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium sulfate, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
7783-20-2
H8N2O4S
132.14
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
7783-20-2
H8N2O4S
132.14
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
Assay Percent Range 99.5%
Linear Formula (NH4)2SO4
Merck Index 15,552
Solubility Information Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
Formula Weight 132.14
Percent Purity 99.5%
Grade Analysis
Chloride 3ppm max
Density 1.76
Packaging Fibre drum
Phosphate 5ppm max
Residue after Ignition 0.01% max. (as SO4)
Solubility Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
Melting Point 280.0°C
Quantity 25kg
Chemical Name or Material Ammonium sulfate
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 231-984-1