Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium sulfate, 99.9995%, (trace metal basis), Acros Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
7783-20-2
H8N2O4S
132.14
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
7783-20-2
H8N2O4S
132.14
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
For ultra trace analysis
Assay Percent Range 99.9995%
Linear Formula (NH4)2SO4
Merck Index 15,552
Solubility Information Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
Formula Weight 132.14
Physical Form Powder or Crystals
Percent Purity 99.9995%
Grade Trace Metal Basis
Gravimetry 23.7 to 24.9 % (S)
Packaging Glass bottle
Solubility Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
Total Trace Metal Impurities 5ppm max.
Color Yellow to Orange
Melting Point 280.0°C
Quantity 250g
Chemical Name or Material Ammonium sulfate
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-984-1