Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium sulfate, ACS, 99.0% min, Alfa Aesar™

Click to view available options
Quantity:
100g
500g
2kg
10kg
7783-20-2
H8N2O4S
132.134
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
7783-20-2
H8N2O4S
132.134
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
Assay Percent Range 99.0%
Solubility Information Highly soluble in water. It is insoluble in acetone,ether and ethanol. It is also insoluble in liquid ammonia.
Formula Weight 132.14
Physical Form Granular
Grade ACS
Density 1.769 g/mL
Color White
Melting Point ∽280°C (decomposition)
Quantity 100g
Chemical Name or Material Ammonium sulfate, For analysis ACS

H303

EINECSNumber : 231-984-1

RTECSNumber : BS4500000

TSCA : Yes

Recommended Storage : Ambient temperatures