Ammonium sulfate, ACS, 99.0% min, Alfa Aesar

Manufacturer: Alfa Aesar 11566A3

Quantity 2kg

 View more versions of this product

Catalog No. AA11566A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-20-2
    Physical Form Granular
    Assay Percent Range 99% min.
    Molecular Formula H8N2O4S
    Formula Weight 132.14
    MDL Number MFCD00003391
    Chemical Name or Material Ammonium sulfate
    Grade ACS Grade

    Specifications

    Formula Weight 132.14
    Quantity 2kg
    Color White
    EINECS Number 231-984-1
    Physical Form Granular
    Assay Percent Range 99% min.
    Molecular Formula H8N2O4S
    Appearance White granular
    MDL Number MFCD00003391
    CAS 7783-20-2
    Health Hazard 1 H303
    Chemical Name or Material Ammonium sulfate, For analysis ACS
    Grade ACS Grade

    H303

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit