Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, ACS, ≥99%, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
7783-20-2
H8N2O4S
132.134
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
7783-20-2
H8N2O4S
132.134
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]

Used in purification of enzymes and other proteins by precipitation

Linear Formula (NH4)2SO4
Formula Weight 132.14
Physical Form Powder
Percent Purity ≥99%
Grade ACS
Packaging Plastic bottle
Specific Gravity 1.769
Melting Point ~280°C (decomposition)
Quantity 5kg
Chemical Name or Material Ammonium Sulfate