Ammonium Sulfate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker 079201

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Catalog No. 02-002-329

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7783-20-2
    Molecular Formula (NH4)2SO4
    Formula Weight 132.14
    Chemical Name or Material Ammonium Sulfate
    Grade Baker Analyzed™ ACS Reagent

    Specifications

    Formula Weight 132.14
    Packaging Poly Bottle
    Physical Form Granular
    Specific Gravity 1L = 1.77kg
    Molecular Formula (NH4)2SO4
    CAS 7783-20-2
    Quantity 500g
    Chemical Name or Material Ammonium Sulfate
    Grade Baker Analyzed™ ACS Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit