Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, Granular, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
100 lbs.
12 kg
2.5 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7783-20-2
H8N2O4S
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
N.N.OS(O)(=O)=O
7783-20-2
H8N2O4S
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
N.N.OS(O)(=O)=O

AM185, 7783-20-2, (NH4)2SO4

Spectrum™ Ammonium Sulfate, Granular, FCC is used as an acidity regulator in flours and breads. It is listed on the FDA′s Generally Recognized As Safe (GRAS) list of substances. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Percent Purity 99 to 100.5%
Ignition Residue 0.25%
Lead (Pb) 3 mg/kg
Packaging Fiber Drum
Selenium (Se) 0.003%
Quantity 100 lbs.