Ammonium Sulfate (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 3kg

 View more versions of this product

Catalog No. A701-3

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7783-20-2
    Physical Form Solid
    Molecular Formula H8N2O4S
    Formula Weight 132.14
    MDL Number MFCD00003391
    Chemical Name or Material Ammonium Sulfate
    Grade Laboratory Grade

    Specifications

    Melting Point 280°C
    Quantity 3kg
    Formula Weight 132.14
    Density 1.77g/mL
    Physical Form Solid
    Color Colorless
    pH 5 to 6
    Molecular Formula H8N2O4S
    Health Hazard 3 Emergency Overview
    May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

    NFPA

    Health:1
    Flammability:0
    Instability:0
    Identification Pass Test
    Packaging Glass Bottle
    MDL Number MFCD00003391
    CAS 7783-20-2
    ChemAlert Storage Symbol Gray
    Health Hazard 2 CAUTION!
    Chemical Name or Material Ammonium Sulfate
    Grade Laboratory Grade
     

    CAUTION!

    Emergency Overview
    May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

    NFPA

    Health:1
    Flammability:0
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit