Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfate, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Poly Bottle
Poly Pail
7783-20-2
H8N2O4S
132.134
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
7783-20-2
H8N2O4S
132.134
BFNBIHQBYMNNAN-UHFFFAOYSA-N
ammonium sulfate,diammonium sulfate,ammonium sulphate,mascagnite,sulfuric acid diammonium salt,ammonium sulfate 2:1,ammoniumsulfate,actamaster,sulfuric acid, diammonium salt,diammonium sulphate
6097028
CHEBI:62946
diazanium;sulfate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Protein Precipitation, Liquid Chromatography and Molecular Biology Applications
Formula Weight 132.14
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Specific Gravity 1L = 1.77kg
Quantity 1kg
Chemical Name or Material Ammonium Sulfate