Ammonium Sulfide Solution (Certified), Fisher Chemical

Packaging HDPE
Quantity 500mL

 View more versions of this product

Catalog No. A705-500

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 12135-76-1
    Assay Percent Range 20 to 24 %
    MDL Number MFCD00010892
    Chemical Name or Material Ammonium Sulfide Solution
    Assay 20 to 24%
    Grade Certified Grade

    Specifications

    Quantity 500mL
    DOT Information DOT Class 8, : Corrosive
    Viscosity [NULL]
    Color Yellow
    Physical Form Liquid
    Density >1
    Vapor Pressure 450mmHg at 20°C
    Molecular Formula H8N2S
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. Causes burns by all exposure routes. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

    NFPA

    Health:3
    Flammability:2
    Instability:1
    Assay Percent Range 20 to 24 %
    Ignition Residue 0.05% max.
    Additional Information Vapor Pressure: 450 mmHg (20°C)
    Identification Pass Test
    Arsenic (As) 3ppm max.
    Carbonate Pass Test
    Chloride Pass Test
    MDL Number MFCD00010892
    CAS 12135-76-1
    UN Number 2683
    Health Hazard 2 DANGER!
    Packaging HDPE
    Chemical Name or Material Ammonium Sulfide Solution
    Assay 20 to 24%
    Grade Certified Grade

    DANGER!

    Emergency Overview
    Flammable liquid and vapor. Causes burns by all exposure routes. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

    NFPA

    Health:3
    Flammability:2
    Instability:1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit