Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfite, Crystal, Purified, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
100 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7783-11-1
7783-11-1
Grade Purified
CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.