Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Tartrate, Crystal, 97%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3164-29-2
3164-29-2
Grade Reagent
CAS Max % 1
Percent Purity 0.97
Chloride 5e-05
Heavy Metals 1e-05
Iron (Fe) 1e-05
Packaging Poly Bottle
Residue after Ignition 0.0001
Sulfate 5e-05
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.