Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Ammonium Tartrate Dibasic, purum p.a., ≥99.0% (T), Honeywell™

Click to view available options
Quantity:
250g
1kg
Packaging:
Poly Bottle
3164-29-2
C4H12N2O6
184.148
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate,diammonium l-+-tartrate,l-tartaric acid, ammonium salt,unii-ji8ud88354,diammonium l-tartrate,r-2,3-dihydroxybutanedioic acid diammonium salt,ammonium tartrate dibasic,butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt,ammonium l-+-tartrate,diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
diazanium;(2R,3R)-2,3-dihydroxybutanedioate
C(C(C(=O)[O-])O)(C(=O)[O-])O.[NH4+].[NH4+]
3164-29-2
C4H12N2O6
184.148
MFCD00013073
NGPGDYLVALNKEG-OLXYHTOASA-N
ammonium d-tartrate,diammonium l-+-tartrate,l-tartaric acid, ammonium salt,unii-ji8ud88354,diammonium l-tartrate,r-2,3-dihydroxybutanedioic acid diammonium salt,ammonium tartrate dibasic,butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt,ammonium l-+-tartrate,diammonium 2r,3r-2,3-dihydroxysuccinate
13652278
CHEBI:63075
diazanium;(2R,3R)-2,3-dihydroxybutanedioate
C(C(C(=O)[O-])O)(C(=O)[O-])O.[NH4+].[NH4+]

Purum p.a., crystallized,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Linear Formula (NH4)2C4H4O6
Beilstein 6120352
Appearance Crystals
Formula Weight 184.15g/mol
Physical Form Crystallized
Percent Purity ≥99.0% (T)
Grade purum p.a.
Density 1.601g/cu.cm
Packaging Poly Bottle
Residue after Ignition ≤0.05% (as SO4)
Color White
Quantity 1kg
Chemical Name or Material Ammonium Tartrate Dibasic
Show More Show Less

EINECSNumber : 221-618-9