Alfa Aesar™ Ammonium tellurate, 99.5%

Manufacturer: Alfa Aesar™ 1448709

Quantity 10g

 View more versions of this product

Catalog No. AA1448709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13453-06-0
    Formula Weight 227.67
    MDL Number MFCD00015964
    UN Number UN3284
    Chemical Name or Material Ammonium tellurate

    Specifications

    Formula Weight 227.67
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00015964
    CAS 13453-06-0
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.
    RTECS Number WY2415000
    TSCA TSCA
    Mesh Size −60 Mesh
    Physical Form Powder
    UN Number UN3284
    Color White
    Quantity 10g
    Chemical Name or Material Ammonium tellurate
    Assay 99.5%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit