Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium tetrachloropalladate(II), Pd 36% min

Manufacturer:  Alfa Aesar™ 1188206

Quantity 5g

 View more versions of this product

Catalog No. AA1188206


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13820-40-1
284.31
Ammonium chloropalladite
(NH4)2PdCl4
MFCD00010884
Ammonium tetrachloropalladate(II)

Specifications

13820-40-1
(NH4)2PdCl4
MFCD00010884
Pd 36% min.
Hygroscopic
5g
Powder
284.31
Ammonium chloropalladite
2.17g/mL
Dark green to brown
Ammonium tetrachloropalladate(II)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P310-P362-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
mmediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BP5400000

TSCA : TSCA

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit