Alfa Aesar™ Ammonium tetrachloropalladate(II), Premion™, 99.998% (metals basis), Pd 37%

Manufacturer: Alfa Aesar™ 1082414

Quantity 25g

 View more versions of this product

Catalog No. AA1082414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13820-40-1
    Linear Formula (NH4)2PdCl4
    Formula Weight 284.31
    MDL Number MFCD00010884
    Chemical Name or Material Ammonium tetrachloropalladate(II)
    Grade Premion™

    Specifications

    Formula Weight 284.31
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310-P362-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    mmediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H318-H302-H315
    Causes serious eye damage.
    Harmful if swallowed.
    Causes skin irritation.
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00010884
    Sensitivity Hygroscopic
    CAS 13820-40-1
    Color Dark green to brown
    Health Hazard 1 Danger
    Density 2.17g/mL
    Linear Formula (NH4)2PdCl4
    Quantity 25g
    Concentration Pd 37%
    RTECS Number BP5400000
    Chemical Name or Material Ammonium tetrachloropalladate(II)
    Assay 99.998%, (metals basis)
    Grade Premion™

    GHS H Statement
    H318-H302-H315
    Causes serious eye damage.
    Harmful if swallowed.
    Causes skin irritation.

    GHS P Statement
    P280-P305+P351+P338-P310-P362-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    mmediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit