Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tetrachloroplatinate(II), 51 to 54% Pt, ACROS Organics™

Click to view available options
Quantity:
1g
CAS 13820-41-2
Molecular Formula H8Cl4N2Pt
Linear Formula (NH4)2PtCl4
MDL Number MFCD00010885
Merck Index 15, 546
Synonym ammonium tetrachloroplatinate ii,diammonium tetrachloroplatinate,ammonium platinous chloride,diammonium tetrachloroplatinate 2-,ammonium tetrachloroplatinate,bis ammonium tetrachloroplatinate 2-,ammonium chloroplatinate ii,unii-z5ny20a9s2,h8cl4n2pt,epitope id:151521
Formula Weight 372.97
Physical Form Crystalline Powder
Percent Purity 51 to 54% (Pt)
Density 2.93g/mL
Packaging Glass bottle
Color Brown-Orange to Red
Quantity 1g
Chemical Name or Material Ammonium tetrachloroplatinate(II), 51 to 54% Pt
Show More Show Less

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 237-499-1

RTECSNumber : TP1840000

TSCA : TSCA