Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tetrachloroplatinate(II), 99.998%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13820-41-2
H8Cl4N2Pt
ammonium tetrachloroplatinate ii,diammonium tetrachloroplatinate,ammonium platinous chloride,diammonium tetrachloroplatinate 2-,ammonium tetrachloroplatinate,bis ammonium tetrachloroplatinate 2-,ammonium chloroplatinate ii,unii-z5ny20a9s2,h8cl4n2pt,epitope id:151521
13820-41-2
H8Cl4N2Pt
ammonium tetrachloroplatinate ii,diammonium tetrachloroplatinate,ammonium platinous chloride,diammonium tetrachloroplatinate 2-,ammonium tetrachloroplatinate,bis ammonium tetrachloroplatinate 2-,ammonium chloroplatinate ii,unii-z5ny20a9s2,h8cl4n2pt,epitope id:151521
For ultra trace analysis
Assay Percent Range 99.998%
Linear Formula (NH4)2PtCl4
Formula Weight 372.97
Physical Form Crystals
Percent Purity 51.5 to 53.1% (Pt)
Grade Trace Metal Basis
Density 2.9300g/mL
Packaging Glass bottle
Total Trace Metal Impurities 20ppm max.
Color Pink
Quantity 5g
Chemical Name or Material Ammonium tetrachloroplatinate(II)
Show More Show Less

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger