Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tetrathiomolybdate, 99.95%, (trace metal basis), Acros Organics

Click to view available options
Quantity:
1g
10g
50g
250g
Packaging:
Glass bottle
15060-55-6
H8MoN2S4
260.28
ZKKLPDLKUGTPME-UHFFFAOYSA-N
ammonium tetrathiomolybdate,molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4,tiomolibdate diammonium,unii-4v6i63lw1e,ammonium thiomolybdate,molybdate 2-, tetrathioxo-, diammonium, t-4,ammonium tetrathiomolybdate vi,molybdate 2-, tetrathioxo-, diammonium, beta-4
15251598
diazanium;bis(sulfanylidene)molybdenum;sulfanide
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
15060-55-6
H8MoN2S4
260.28
ZKKLPDLKUGTPME-UHFFFAOYSA-N
ammonium tetrathiomolybdate,molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4,tiomolibdate diammonium,unii-4v6i63lw1e,ammonium thiomolybdate,molybdate 2-, tetrathioxo-, diammonium, t-4,ammonium tetrathiomolybdate vi,molybdate 2-, tetrathioxo-, diammonium, beta-4
15251598
diazanium;bis(sulfanylidene)molybdenum;sulfanide
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
For ultra trace analysis
Assay Percent Range 99.95%
Linear Formula (NH4)2MoS4
Merck Index 15,557
Formula Weight 260.28
Physical Form Powder or Cyrstals
Percent Purity 99.95%
Grade Trace Metal Basis
Gravimetry 35.5 to 38.1% (Mo)
Molybdenum (Mo) 35.5 to 38.1%
Packaging Glass bottle
Total Trace Metal Impurities 500ppm max.
Color White
Melting Point 300.0°C
Quantity 250g
Chemical Name or Material Ammonium tetrathiomolybdate
Show More Show Less

GHS Signal Word: Warning