Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Thiocyanate, 0.1 N Solution, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
1762-95-4,7732-18-5
1762-95-4,7732-18-5
Grade Chemical Solution
Concentration 0.1 N ± 0.005 N
Packaging Amber Glass Bottle
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.