Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium thiocyanate, 99+%, extra pure, ACROS Organics™
Click to view available options
Quantity:
500g
1kg
2.5kg
10kg
Packaging:
Plastic Drum
Plastic bottle
Specifications
Assay Percent Range | 99% min. (on dried basis) (Argentometry) |
Linear Formula | NH4SCN |
Merck Index | 15,558 |
Solubility Information | Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol,acetone and ammonia,practically insoluble in chcl3 and ethyl acetate |
Formula Weight | 76.12 |
Physical Form | Crystals |
Percent Purity | 99+% |
Grade | Extra Pure |
Density | 1.3000g/mL |
Iron (Fe) | 2ppm max. |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF
GHS Signal Word: Danger
EINECSNumber : 217-175-6