Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Thiocyanate, Crystal, Reagent, ACS, 97.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1762-95-4
CH4N2S
76.12
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium cyanosulfanide
[NH4+].[S-]C#N
1762-95-4
CH4N2S
76.12
SOIFLUNRINLCBN-UHFFFAOYSA-N
ammonium cyanosulfanide
[NH4+].[S-]C#N

A1270, 1762-95-4, NH4SCN

Spectrum™ Ammonium Thiocyanate, Crystal, Reagent, ACS is an inorganic laboratory reagent commonly used by the soft drinks industry to determine the iron content in soft drinks by colorimetry. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Packaging Amber Glass Bottle
Quantity 125 g
CAS Max % 100%
Percent Purity 97.5%