Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Thiocyanate, Crystal, Reagent, ACS, 97.5%, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Ammonium Thiocyanate, Crystal, Reagent, ACS is an inorganic laboratory reagent commonly used by the soft drinks industry to determine the iron content in soft drinks by colorimetry. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 6 x 500 g |
Percent Purity | 97.5% |
Grade | ACS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction