Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog Number AAH2657722
Click to view available options
Quantity:
25 g
100 g
3336-58-1
C2H5F3NO2
132.06
MFCD00012615,MFCD03095537
YCNIBOIOWCTRCL-UHFFFAOYSA-O
ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
86601334
azanium;2,2,2-trifluoroacetic acid
[NH4+].OC(=O)C(F)(F)F
3336-58-1
C2H5F3NO2
132.06
MFCD00012615,MFCD03095537
YCNIBOIOWCTRCL-UHFFFAOYSA-O
ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
86601334
azanium;2,2,2-trifluoroacetic acid
[NH4+].OC(=O)C(F)(F)F

Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

Solubility
Soluble in Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture. Keep under dry inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 123°C to 126°C
Linear Formula CF3COONH4
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in Water.
Formula Weight 131.05
Percent Purity 98%
Chemical Name or Material Ammonium trifluoroacetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-072-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit