Alfa Aesar™ Ammonium tungsten oxide, 99.99+% (metals basis)

Manufacturer: Alfa Aesar™ 4458830

Quantity 250g

 View more versions of this product

Catalog No. AA4458830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11140-77-5
    Molecular Formula (NH4)2WO4
    Formula Weight 283.91
    MDL Number MFCD00015956
    Chemical Name or Material Ammonium tungsten oxide

    Specifications

    Formula Weight 283.91
    Physical Form Powder
    Molecular Formula (NH4)2WO4
    CAS 11140-77-5
    Quantity 250g
    Solubility Information Soluble in warm water
    MDL Number MFCD00015956
    Chemical Name or Material Ammonium tungsten oxide
    Assay ≥99.99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit