Alfa Aesar™ Ammonium tungsten oxide hydrate

Manufacturer: Alfa Aesar™ 2264014

Quantity 25g

 View more versions of this product

Catalog No. AA2264014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11120-25-5
    Linear Formula (NH4)10W12O41·xH2O
    Formula Weight 3042.56 (Anhydrous)
    MDL Number MFCD00015956
    Synonym Ammonium paratungstate; Ammonium tungstate
    Chemical Name or Material Ammonium tungsten oxide hydrate

    Specifications

    Formula Weight 3042.56 (Anhydrous)
    Odor Odorless
    Synonym Ammonium paratungstate; Ammonium tungstate
    Quantity 25g
    Solubility Information Soluble in water; Insoluble in alcohol
    Density 2.3g/mL
    TSCA TSCA
    Physical Form Powder
    Linear Formula (NH4)10W12O41·xH2O
    MDL Number MFCD00015956
    CAS 11120-25-5
    Color White
    Chemical Name or Material Ammonium tungsten oxide hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit