Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium tungsten oxide pentahydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1089922

Quantity 100g

 View more versions of this product

Catalog No. AA1089922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1311-93-9
99.999% (Metal basis)
3132.64
Ammonium tungsten oxide pentahydrate
Crystalline Powder
(NH4)10W12O41·5H2O
MFCD00015956

Specifications

1311-93-9
99.999% (Metal basis)
White crystalline
MFCD00015956
White
Ammonium tungsten oxide pentahydrate
Crystalline Powder
(NH4)10W12O41·5H2O
3132.64
Odorless
100g

Safety and Handling

SDS & Safety Documents

EINECSNumber : 234-364-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit