Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Vanadate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle

A-314, 7803-55-6

Spectrum™ Ammonium Vanadate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 7803-55-6,7732-18-5
CAS Max % 0.1%,99.9%
Molecular Formula H4NO3V
UN Number UN2031
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILES N.O[V](=O)=O
IUPAC Name vanadiumoylol amine
Molecular Weight (g/mol) 116.98
Packaging Poly Bottle
Quantity 500 mL