Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amodiaquine Mouse anti-Non-species specific, Clone: 6D10, Novus Biologicals™
Mouse Monoclonal Antibody
$378.00
Specifications
Antigen | Amodiaquine |
---|---|
Clone | 6D10 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:4000 |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105106
|
Novus Biologicals
NBP105106 |
100 μg |
Each of 1 for $378.00
|
|
Description
Amodiaquine Monoclonal specifically detects Amodiaquine in Non-species specific samples. It is validated for ELISA.Specifications
Amodiaquine | |
1.0 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Nonspecies specific | |
IgG2a κ | |
Protein A or G purified | |
Specific for the non-quinoline sidechain of amodiaquine. Can be used for quantitation of amodiaquine in human serum or plasma with a detection limit of app. 2 ng/ml. |
6D10 | |
ELISA 1:4000 | |
Monoclonal | |
Purified | |
RUO | |
4-amino-2(diethylaminomethyl)phenol a preparation of a carrier protein | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title