Learn More
Abnova™ AMOTL1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00154810T01
Catalog No. 89-933-320
Add to cart
Description
Description
Specifications
Specifications
293T
NM_130847.2
pCMV-AMOTL1 full-length
100 μL
Human
AMOTL1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 106.6kDa
Transient overexpression cell lysate was tested with anti-AMOTL1 antibody (Mfr. No. H00154810-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot