Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Amoxicillin clavulanate, 100mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
25mL
50mL
MFCD28100498
Augmentin
MFCD28100498
Augmentin
Physical Form Liquid
Quantity 25mL
Chemical Name or Material Amoxicillin clavulanate, 100mg/mL in distilled water, sterile-filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : -30° to -10°C