Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Amoxicillin sodium salt, 100mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
10mL
25mL
34642-77-8
C16H18N3NaO5S
387.386
MFCD08063910
BYHDFCISJXIVBV-ZONSQSGVSA-M
131632470
sodium;(2S,5R,6S)-6-[[2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)[O-])C.[Na+]
34642-77-8
C16H18N3NaO5S
387.386
MFCD08063910
BYHDFCISJXIVBV-ZONSQSGVSA-M
131632470
sodium;(2S,5R,6S)-6-[[2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)[O-])C.[Na+]
Physical Form Liquid
Quantity 10mL
Chemical Name or Material Amoxicillin sodium salt, 100mg/mL in distilled water, sterile-filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XH8300000

storageNote1 : -30° to -10°C