Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampac™ Flexibles KeepSafe™ Returnable Polymailer Bags

Flexible packaging alternative to corrugated box. Ampac Flexibles Returnable Polymailer Bags with dual permanent adhesive closure offers better protection from the elements.

$113.20 - $161.50

Specifications

Type Returnable Polymailer
Material Tri-layer LLDPE Film
Applications Shipping Package
Closure Type 2X Permanent Adhesive
Color White/Silver
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
01-813-000
View Documents
Ampac™
PRM1215
15.5 x 12 in. 500/Cs. Case of 500 for $161.50
Add to cart
 
01-813-001
View Documents
Ampac™
PRM1419
19 x 14.5 in. 250/Cs. Case of 250 for $113.20
Add to cart
 
Description

Description

  • Flexible packaging unlike to that of conjuagted box
  • Constructed of coextruded tri-layer LLDPE film
  • 2X permanent adhesive closures
  • 2x anti-static release liners
  • Upto 30% recycled content
Specifications

Specifications

Returnable Polymailer
Shipping Package
White/Silver
Lip: 3 in.
Tri-layer LLDPE Film
2X Permanent Adhesive
Up to 30% recycled content
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit