Learn More
Ampac™ Flexibles StandPAK Pouches Available on GSA/VA Contract for Federal Government customers only.
Heat sealable 4.5mL stand-up pouch with tear notches
$907.50
Specifications
Capacity (Metric) 4.5mL
View More Specs
Products 1
Catalog Number Mfr. No. Material Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Material Dimensions (L x W) Price Quantity & Availability  
01-812-74 Available on GSA/VA Contract for Federal Government customers only. View Documents Ampac™
SP6801EZ500
Polyester 9 x 6 in. (22.9 x 58.1cm) Case of 500 for $907.50
Add to cart
Description
Description
  • Perfect for medical, environmental, and industrial purposes
  • Withstand boiling and freezing
  • Can be used for long-term specimen/evidence storage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
4.5mL