Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | AMPD2 |
---|---|
Dilution | Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AMPD2 Polyclonal antibody specifically detects AMPD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
AMPD2 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
metabolism | |
PBS (pH 7.2), 40% Glycerol | |
271 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
adenosine monophosphate deaminase 2, adenosine monophosphate deaminase 2 (isoform L), AMP deaminase 2, AMP deaminase isoform L, AMPD, EC 3.5.4.6 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RRKGLDVAEPGPSRCRSDSPAVAAVVPAMASYPSGSGKPKAKYPFKKRASLQASTAAPEARGGLGAPPLQSARSLPGPAPCLKHFPL | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title