Learn More
Aviva Systems Biology Corp AMPH Antibody - N-terminal region (ARP55434_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMPH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AMPH. Alternate names: AMPH1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61507