Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amphiregulin Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen Amphiregulin
Concentration 0.21 mg/mL
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-0665
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
160361AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

The protein encoded by this gene is a member of the epidermal growth factor family. It is an autocrine growth factor as well as a mitogen for astrocytes, Schwann cells, and fibroblasts. It is related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). This protein interacts with the EGF/TGF-alpha receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. This encoded protein is associated with a psoriasis-like skin phenotype.
Specifications

Specifications

Amphiregulin
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Human
IgG
-20°C
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit