Learn More
Gemini Bio Products AMPHOTERICIN B SOLUTION 100ML
SDP
Supplier:  Gemini Bio Products 400104/100
Encompass_Preferred
Amphotericin B is one of the earliest compounds designed to kill fungi. It acts by forming channels in fungal membranes, causing the cells to become leaky. It is, of course, toxic to eukaryotic cells in culture.
Catalog No. 50-753-3017
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.