Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ampicillin, Ready-to-Use aqueous soln.,/mL, 0.2 micron filtered

Click to view available options
Quantity:
10mL
25mL
69-53-4
C16H19N3O4S
349.405
AVKUERGKIZMTKX-NJBDSQKTSA-N
6249
CHEBI:28971
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C
69-53-4
C16H19N3O4S
349.405
AVKUERGKIZMTKX-NJBDSQKTSA-N
6249
CHEBI:28971
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C
Odor Odorless
Physical Form Liquid
Color Yellow
Quantity 25mL
Sensitivity Light Sensitive
Chemical Name or Material Ampicillin, eady-to-Use aqueous soln., 100mg/mL, 0.2 micron filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XH8400000

storageNote1 : -30° to -10°C