Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ampicillin Sodium Salt, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
69-52-3
C16H21N3NaO4S
374.411
MFCD00064313
BSFVNXCYXDYHOD-ZQDFAFASSA-N
ampicillin sodium
131673879
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]
69-52-3
C16H21N3NaO4S
374.411
MFCD00064313
BSFVNXCYXDYHOD-ZQDFAFASSA-N
ampicillin sodium
131673879
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]
Formula Weight 371.39
Physical Form Powder
Grade Ultrapure
Packaging Plastic bottle
Melting Point 215°C
Quantity 25g
Chemical Name or Material Ampicillin Sodium Salt