Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin, Trihydrate, Micronized, USP, 900-100.5 μg/mg, Spectrum™

Click to view available options
Quantity:
5 kg
20 kg
Packaging:
Poly Pail
7177-48-2
C16H25N3O7S
403.45
RXDALBZNGVATNY-CWLIKTDRSA-N
(2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
7177-48-2
C16H25N3O7S
403.45
RXDALBZNGVATNY-CWLIKTDRSA-N
(2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O

A1819, 7177-48-2, C16H19N3O4S•3H2O

Spectrum™ Ampicillin, Trihydrate, Micronized, USP is an antibiotic used to treat a range of bacterial infections. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Water 12.0-15%
Quantity 20 kg
CAS Max % 100%