Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 1 Mouse anti-Human, Mouse, Rat, Primate, Allophycocyanin/Cy7, Clone: 2B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP222127APCCY7

Catalog No. N222127APC7


Add to cart

Description

Description

AMPK alpha 1 Monoclonal antibody specifically detects AMPK alpha 1 in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry.
Specifications

Specifications

AMPK alpha 1
2B7
Flow Cytometry
Conjugated
PRKAA1
Purified recombinant fragment of human AMPK alpha 1 expressed in E. coli. [UniProt No. Q13131]
Ammonium sulfate precipitation
Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control
Primary
5562.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5′-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5′-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse, Primate, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit