Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha Rabbit anti-Human,Mouse,Rat, KHL, Clone: 2D22, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen AMPK alpha
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB110025MI
View Documents
MilliporeSigma
ZRB110025UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11004XMI
View Documents
MilliporeSigma
ZRB11004X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

AMPK alpha Monoclonal antibody specifically detects AMPK alpha in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

AMPK alpha
KHL
Rabbit
5'-AMP-activated protein kinase catalytic subunit alpha-1;AMPK subunit alpha-1;Acetyl-CoA carboxylase kinase;ACACA kinase;Hydroxymethylglutaryl-CoA reductase kinase;HMGCR kinase;Tau-protein kinase PRKAA1
KLH-conjugated liear peptide corresponding to 17 amino acids from the C-terminal half of human AMPK alpha, isoform 1.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat
PRKAA1, AMPK1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit